

Sodium methoxide ≥96.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:124-41-4
:
Formula:CH₃NaO
MDL Number:MFCD00012179
:54.02 g/mol
Merck Index:14,08643
Melting Pt:127 °C
Density:1,3 g/cm³ (20 °C)
EINECS:204-699-5
UN:1431
ADR:4.2,II
:GHS02
:GHS05
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 96.0 %
Specification Test Results
Related Information
Frequently Bought Together