Order Entry
Belgium
ContactUsLinkComponent
 
 
Lactic acid ≥85.0% (by titrimetric analysis)
 :  TCI
 
undefined
 
Lactic acid ≥85.0% (by titrimetric analysis)
 :  TCI

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (Neutralization back titration):
    min. 85.0 %

 

Frequently Bought Together