Order Entry
Belgium
ContactUsLinkComponent
α,α,α-Trifluoro-o-tolyl isocyanate ≥98.0% (by GC)
α,α,α-Trifluoro-o-tolyl isocyanate ≥98.0% (by GC)
Catalog # TCIAI0412-10G
Supplier:  TCI
CAS Number:  
undefined
α,α,α-Trifluoro-o-tolyl isocyanate ≥98.0% (by GC)
Catalog # TCIAI0412-10G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :187.12 g/mol
Boiling Pt:79°C
Flash Pt:58°C
Storage Temperature:Refrigerator
EINECS:218-925-5
UN:2285
ADR:6.1,II
 :GHS02
 :GHS06
 :GHS08
CAS Number:2285-12-3
Formula:C₈H₄F₃NO
MDL Number:MFCD00002008