Order Entry
Belgium
ContactUsLinkComponent
 
3-Pentyn-1-ol ≥98.0% (by GC)
  TCIAP0816-5ML
 :  TCI
 :  
undefined
3-Pentyn-1-ol ≥98.0% (by GC)
  TCIAP0816-5ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210
  • Environmentally Preferable:

 

 

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 98.0 %

 

 

 : Pent-3-yn-1-ol
 :
 :84.12 g/mol
 :233-550-7
 :1987
 :GHS02
 :GHS07
 :Refrigerator
 :10229-10-4
 :C₅H₈O
 :MFCD00002956
 :154°C
 :54°C
 :3 III