1-Hexanesulphonic acid sodium salt ≥98.0% (by titrimetric analysis)
: TCI
:2832-45-3
:MFCD00007542
:C₆H₁₃O₃SNa
:300 °C
:188.22 g/mol
:Ambient
:220-601-3
- Appearance:White to Almost white powder to crystal
- Purity (Ion exchange titration):min. 98.0 %
- Solubility in Water:almost transparency
Frequently Bought Together