


p-Phenylenediamine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:106-50-3
:
Formula:C₆H₈N₂
MDL Number:MFCD00007901
:108.14 g/mol
Merck Index:14,07285
Boiling Pt:267°C
Melting Pt:139…141 °C
Density:1,251 g/cm³ (20 °C)
Flash Pt:156 °C
Storage Temperature:Ambient
EINECS:203-404-7
UN:1673
ADR:6.1,III
:GHS06
:GHS08
:GHS09
Specification Test Results
- Appearance:White to Light yellow to Light red powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:140.0 to 145.0 deg-C
- Solubility in hot EtOH (95vol%):almost transparency
Specification Test Results
Frequently Bought Together