
Diethyl phthalate ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:84-66-2
:
Formula:C₁₂H₁₄O₄
MDL Number:MFCD00009111
:222.24 g/mol
Merck Index:14,07371
Boiling Pt:296°C
Melting Pt:–41°C
Density:1,12 g/cm³ (20 °C)
Flash Pt:162°C (closed cup)
EINECS:201-550-6
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless clear liquid
- Purity (GC):min. 99.0 %
Specification Test Results
Frequently Bought Together