Order Entry
Belgium
ContactUsLinkComponent
L(+)-Glutamine ≥99.0% (by titrimetric analysis)
L(+)-Glutamine ≥99.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
L(+)-Glutamine ≥99.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Optical purity (LC):
    min. 98.0 ee%
  • Purity (Nonaqueous Titration):
    min. 99.0 %
  • Solubility in Water:
    almost transparency

Specification Test Results

Related Information

Frequently Bought Together