L(+)-Asparagine monohydrate ≥99.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
Synonyms: L-Asparagine monohydrate, H-Asn-OH·H₂O, (S)-(+)-Asparagine monohydrate, (S)-2,4-Diamino-4-oxobutanoic acid hydrate, H-Asn-OH.H2O
CAS Number:5794-13-8
:
Formula:C₄H₈N₂O₃·1H₂O
MDL Number:MFCD00151038
:150.13 g/mol
Merck Index:14,00837
Melting Pt:215…217 °C
Density:1,567 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-735-9
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White powder to crystal
- Purity (HPLC):min. 99.0 area%
- Optical purity (LC):min. 98.0 ee%
- Purity (Nonaqueous Titration):min. 99.0 % (calcd.on dried substance)
- Drying loss:11.5 to 12.5 %
Specification Test Results
Frequently Bought Together