L(+)-Aspartic acid ≥99.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:56-84-8
:
Formula:C₄H₇NO₄
MDL Number:MFCD00002616
:133.1 g/mol
Merck Index:14,00840
Boiling Pt:311 °C (1013 hPa)
Melting Pt:251 °C
Density:1,6613 g/cm³ (13 °C)
Storage Temperature:Ambient
EINECS:200-291-6
Specification Test Results
- Appearance:White powder to crystal
- Purity (Nonaqueous Titration):min. 99.0 %
- Specific rotation [a]20/D:+24.0 to +26.0 deg (C=8, HCl (1+1))
- Solubility in HCl (1+10):almost transparency
Specification Test Results
Related Information
Frequently Bought Together