Order Entry
Belgium
ContactUsLinkComponent
2-Phenylimidazole ≥98.0% (by GC, titration analysis)
2-Phenylimidazole ≥98.0% (by GC, titration analysis)
Catalog # TCIAP0684-100G
Supplier:  TCI
CAS Number:  
undefined
2-Phenylimidazole ≥98.0% (by GC, titration analysis)
Catalog # TCIAP0684-100G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • Seq:
    3401
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystaline
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    145.0 to 150.0 deg-C

Specification Test Results

About this item

Synonyms: 2-Phenyl-1H-imidazole
 :
 :144.18 g/mol
Boiling Pt:340°C
Melting Pt:143…146 °C
Density:0,6 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:211-581-7
 :GHS07
CAS Number:670-96-2
Formula:C₉H₈N₂
MDL Number:MFCD00005186