Order Entry
Belgium
ContactUsLinkComponent
 
Sodium-1-decanesulphonate ≥98.0% (by titrimetric analysis)
 :  TCI
 
undefined
Sodium-1-decanesulphonate ≥98.0% (by titrimetric analysis)
 :  TCI

 

  • Appearance:
    White powder to crystal
  • Purity (Ion exchange titration):
    min. 98.0 %
  • Solubility in Water:
    almost transparency

 

Frequently Bought Together