3-Phenoxytoluene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:3586-14-9
:
Formula:C₁₃H₁₂O
MDL Number:MFCD00008531
:184.24 g/mol
Boiling Pt:272°C
Density:1,051 g/cm³ (25 °C)
EINECS:222-716-4
UN:3082
ADR:9,III
:GHS09
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together