Order Entry
Belgium
ContactUsLinkComponent
 
D-(-)-Mannitol ≥99.0% (by titrimetric analysis)
 :  TCI
 
undefined
D-(-)-Mannitol ≥99.0% (by titrimetric analysis)
 :  TCI

 

  • Appearance:
    White powder to crystal
  • Purity (Iodometric Titration):
    min. 99.0 %
  • Melting point:
    166.0 to 170.0 deg-C
  • Specific rotation [a]20/D:
    +22.0 to +24.0 deg (C=10, sodium tetraborate solution)
  • Solubility in Water:
    almost transparency

 

Frequently Bought Together