

(±)-1-Methoxy-2-propanol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:107-98-2
:
Formula:CH₃CH(OH)CH₂OCH₃
MDL Number:MFCD00004537
Boiling Pt:120°C
Melting Pt:–97 °C
Density:0,92 g/cm³ (20 °C)
Flash Pt:32°C
Storage Temperature:Ambient
EINECS:203-539-1
ADR:3,III
:90.12 g/mol
UN:3092
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together