
2-Fluoro-4-iodopyridine ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:22282-70-8
:
Formula:C₅H₃FIN
MDL Number:MFCD03092926
:222.99 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:56.0 to 60.0 deg-C
Specification Test Results
Frequently Bought Together