Order Entry
Belgium
ContactUsLinkComponent
 
D(+)-Tryptophan ≥98.0% (by HPLC, titration analysis)
 :  TCI
 
undefined
D(+)-Tryptophan ≥98.0% (by HPLC, titration analysis)
 :  TCI

 

  • Environmentally Preferable:

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Specific rotation [a]20/D:
    +29.0 to +33.0 deg (C=1, H2O)

 

Frequently Bought Together