D(-)-Norleucine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
Synonyms: H-D-Nle-OH, (R)-(-)-Norleucine
CAS Number:327-56-0
:
Formula:C₆H₁₃NO₂
MDL Number:MFCD00008099
Merck Index:14,06706
Melting Pt:301 °C
:131.17 g/mol
EINECS:206-320-9
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Specific rotation [a]20/D:-22.0 to -24.0 deg (C=4.5, 6mol/L HCl)
- Solubility in hot Water:almost transparency
Specification Test Results
Related Information
Frequently Bought Together