Order Entry
Belgium
ContactUsLinkComponent
Hexamethylenetetramine ≥99.0% (by GC, titration analysis)
Flammable
Irritant
Hexamethylenetetramine ≥99.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Flammable
Irritant
Hexamethylenetetramine ≥99.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White powder to crystal
  • Purity (GC):
    min. 99.0 %
  • Purity (Nonaqueous Titration):
    min. 99.0 %
  • Solubility in Water:
    almost transparency

Specification Test Results

Frequently Bought Together