Order Entry
Belgium
ContactUsLinkComponent
Iodine concentrate for dilution to 0.05 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Iodine concentrate for dilution to 0.05 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Catalog # 84591.600
Supplier:  VWR Chemicals
CAS Number:  
undefined
Iodine concentrate for dilution to 0.05 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Catalog # 84591.600
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass ampoule
  • Pk:
    30 mL
  • Seq:
    2345

Specifications

Specification Test Results

  • Titer (20°C; real value 0.2 % accuracy):
    0.04998 - 0.05002 mol/l

Specification Test Results

About this item

 :
 :253.81 g/mol
 :Dose for preparation of 1 litre of solution.
Storage Temperature:Ambient
EINECS:231-442-4
CAS Number:7553-56-2
Formula:I₂
MDL Number:MFCD00011355