Order Entry
Belgium
ContactUsLinkComponent
Caffeine ≥98.0% (by HPLC, titration analysis)
Toxic
Health
Caffeine ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
undefined
Toxic
Health
Caffeine ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystaline
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Solubility in hot Methanol:
    almost transparency

Specification Test Results

Related Information

Frequently Bought Together