Hypoxanthine ≥98.0% (by HPLC and UV method)
Supplier: TCI
Certificates
About this item
CAS Number:68-94-0
:
Formula:C₅H₄N₄O
MDL Number:MFCD00005725
:136.11 g/mol
Merck Index:14,04869
Melting Pt:> 310 °C
EINECS:200-697-3
Specification Test Results
- Appearance:White to Light yellow to Light red powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (UV-vis method):98.0 to 104.0 %
- Lambda max.:247.0 to 251.0 nm (pH 7.0 buffer sol.)
- abs-ratio (a/b):1.29 to 1.35 (E250/E260)
Specification Test Results
Frequently Bought Together