Order Entry
Belgium
ContactUsLinkComponent
Phthalic anhydride ≥98.0% (by GC, titration analysis)
Corrosive
Irritant
Health
Phthalic anhydride ≥98.0% (by GC, titration analysis)
Catalog # TCIAP1614-500G
Supplier:  TCI
CAS Number:  
Corrosive
Irritant
Health
Phthalic anhydride ≥98.0% (by GC, titration analysis)
Catalog # TCIAP1614-500G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Morpholine Method):
    min. 98.0 %
  • Melting point:
    130.0 to 134.0 deg-C

Specification Test Results

About this item

 :
 :148.12 g/mol
Merck Index:14,07372
Boiling Pt:284°C
Melting Pt:131 °C
Density:1,545 g/cm³ (20 °C)
Flash Pt:151 °C
EINECS:201-607-5
UN:2214
ADR:8,III
 :GHS05
 :GHS07
 :GHS08
Storage Temperature:Ambient
CAS Number:85-44-9
Formula:C₈H₄O₃
MDL Number:MFCD00005918