Order Entry
Belgium
ContactUsLinkComponent
 
 
AMP (2-Amino-2-methylpropanol) ≥93.0% (by GC, titration analysis)
 :  TCI
 
undefined
 
AMP (2-Amino-2-methylpropanol) ≥93.0% (by GC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Almost white powder to lump
  • Purity (GC):
    min. 93.0 %
  • Purity (Neutralization titration):
    min. 93.0 %

 

Frequently Bought Together