Order Entry
Belgium
ContactUsLinkComponent
 
 
Guanidinium hydrochloride ≥98.0% (by titrimetric analysis)
 :  TCI
 
undefined
 
Guanidinium hydrochloride ≥98.0% (by titrimetric analysis)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    184.0 to 188.0 deg-C
  • Solubility in Water:
    almost transparency

 

Frequently Bought Together