DL-Norleucine ≥98.0% (by titrimetric analysis)
Catalog # TCIAN0302-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Solubility in hot Water:almost transparency
Specification Test Results
About this item
Synonyms: H-DL-Nle-OH, (±)-Norleucine, 2-Aminohexanoic acid
:
:131.17 g/mol
Merck Index:14,06706
Boiling Pt:246 °C (1013 hPa)
Melting Pt:113 °C
Density:1,17 g/cm³ (20 °C)
EINECS:210-462-7
CAS Number:616-06-8
Formula:C₆H₁₃NO₂
MDL Number:MFCD00064422