Benzoyl isothiocyanate ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:532-55-8
:
Formula:C₈H₅NOS
MDL Number:MFCD00004815
:163.2 g/mol
Merck Index:14,01114
Boiling Pt:143°C
Melting Pt:–1 °C
Density:1,197 g/cm³ (16 °C)
Storage Temperature:Ambient
EINECS:208-540-0
UN:2810
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:Light yellow to Brown clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together