Order Entry
Belgium
ContactUsLinkComponent
 
3-Bromopropionyl chloride ≥95.0% (by GC, titration analysis)
  TCIAB2065-25G
 :  TCI
 :  
undefined
3-Bromopropionyl chloride ≥95.0% (by GC, titration analysis)
  TCIAB2065-25G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308
  • Environmentally Preferable:

 

 

  • Appearance:
    Colorless to Red to Green clear liquid
  • Purity (GC):
    min. 95.0 %
  • Purity (Argentmetric Titration):
    min. 95.0 %

 

 

 : 3-Bromopropanoyl chloride
 :
 :171.42 g/mol
 :55 °C (1013 hPa)
 :1,7 g/cm³ (20 °C)
 :79°C
 :Refrigerator
 :239-515-2
 :3265
 :8,II
 :GHS05
 :15486-96-1
 :BrH₂CCH₂COOCl
 :MFCD00000746