Order Entry
Belgium
Orders LinkContactUsLinkComponent
Bromochlorophenol blue

Bromochlorophenol blue

Glass bottle, 1 g
Catalog # TCIAB0574-1G
Supplier:   TCI
Bromochlorophenol blue
Catalog # TCIAB0574-1G
Supplier:  TCI
CAS Number:  2553-71-1
Bromochlorophenol blue
Bromochlorophenol blue

Glass bottle, 1 g

CAS Number:  2553-71-1
Price
24,20 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-bromo-6-chlorophenol] S,S-dioxide
  • CAS Number:2553-71-1
  • Formula:C₁₉H₁₀Br₂Cl₂O₅S
  • MDL Number:MFCD00047291
  • Molecular Weight:581.06 g/mol
  • Boiling Pt:603 °C (1013 hPa)
  • Melting Pt:250…251 °C
  • EINECS:219-861-0
Specifications

Specification Test Results

  • Appearance:
    Light yellow to Brown powder to crystal
  • Solubility in EtOH (50vol%):
    almost transparency
  • Solubility in Sodium hydroxide solution:
    almost transparency
  • Drying loss:
    max. 1.0 %
  • Ignition residue (Sulfate):
    max. 0.5 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g

Specifications