Orders LinkContactUsLinkComponent
Dissolved oxygen electrodes
Supplier:   KNICK
Dissolved oxygen electrodes
Supplier:  KNICK
Certificates
Dissolved oxygen electrodes
Dissolved oxygen electrodes
2 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • Type
    Concentration
    Temp.
    Material
    Connection
    Ø×L
    For
    Availability
    Price
  • KNICSE340
    SE 340, optical oxygen probe, digital, with NTC 22 kOhm temperature sensor
    SE 340, optical oxygen probe, digital, with NTC 22 kOhm temperature sensor
    SE 340, optical, digital, with temperature sensor
    0 - 20 mg/l
    0...+50 °C
    POM
    M12, 8-pin, 1,5 m cable
    12×140 mm
    Water, wastewater
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 410,00 €
    1 410,00 €
    1 * 1 ST
       1 410,00 €
  • 664-0216
    SE 715-MS, galvanic oxygen probe, digital, with NTC 22 kOhm temperature sensor
    SE 715-MS, galvanic oxygen probe, digital, with NTC 22 kOhm temperature sensor
    SE 715/1-MS, digital, with temperature sensor
    0 - 20 mg/l
    −5...+45 °C
    Polysulfone
    Memosens
    12×120 mm
    Water, wastewater, aquariums
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 410,00 €
    1 410,00 €
    1 * 1 ST
       1 410,00 €