Orders LinkContactUsLinkComponent
Racks for cryogenic vials, Nunc™
Racks for cryogenic vials, Nunc™
   Sustainable Options Available
Racks for cryogenic vials, Nunc™
   Sustainable Options Available
Certificates
Racks for cryogenic vials, Nunc™
4 Options Available Below
Limited Time Sale
Select items below are currently on sale
Product Details & Documents
  • Latch rack with standard ANSI microplate footprint
  • Holds 48 internally-threaded CryoTubes™ or Cryobank vials
  • Use pivoting lid on bench or lock it for transport and storage
  • Format facilitates automated picking/placing, capping/decapping, and multi-channel liquid handling
  • Designed for –80°C and liquid nitrogen (vapour phase) storage
Specifications for Product Family
Specifications
  • Catalog No:
  • 479-1149
  • 479-1147
  • 479-1151
  • 479-1148
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Specifications
Product Family Options
Product Information
  • For tubes
    No. of Holes
    Availability
    Price
  • 1 - 2 ml
    48 (6×8)
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 138,00 €
    138,00 €
    Sale
    Valid until 30/06/2026
    SaleValid until 30/06/2026
    1 * 10 ST
       138,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    479-1149
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 2 ml
    48 (6×8)
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 140,00 €
    140,00 €
    Sale
    Valid until 30/06/2026
    SaleValid until 30/06/2026
    1 * 10 ST
       140,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    479-1147
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 3,6 - 5 ml
    48 (6×8)
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 138,00 €
    138,00 €
    Sale
    Valid until 30/06/2026
    SaleValid until 30/06/2026
    1 * 10 ST
       138,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    479-1151
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 5 ml
    48 (6×8)
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 140,00 €
    140,00 €
    Sale
    Valid until 30/06/2026
    SaleValid until 30/06/2026
    1 * 10 ST
       140,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    479-1148
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact