Orders LinkContactUsLinkComponent
Watchmakers' magnifiers
Watchmakers' magnifiers
Supplier:   ESCHENBACH OPTIK
Watchmakers' magnifiers
Supplier:  ESCHENBACH OPTIK
Certificates
Watchmakers' magnifiers
Watchmakers' magnifiers
4 Options Available Below
Product Details & Documents
PXM® lightweight lenses with antistatic and scratch resistant coating. Ideal as a watchmaker's magnifier or gemstone magnifier.
  • Includes vent to prevent condensation
  • Made of flame resistant plastic material
  • Colour: anthracite
Product Family Options
Product Information
  • Magnification
    Ø
    Availability
    Price
  • ESCH11245
    Magnifier with plano-convex lens
    Magnifier with plano-convex lens
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 27,10 €
    27,10 €
    1 * 1 ST
       27,10 €
  • ESCH11247
    Magnifier with plano-convex lens
    Magnifier with plano-convex lens
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 28,40 €
    28,40 €
    1 * 1 ST
       28,40 €
  • ESCH1124110
    Magnifier with aspheric lens
    Magnifier with aspheric lens
    10×
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 32,90 €
    32,90 €
    1 * 1 ST
       32,90 €
  • ESCH1130
    Magnifier with dual lens
    Magnifier with dual lens
    15×
    13 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 48,80 €
    48,80 €
    1 * 1 ST
       48,80 €
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.