Orders LinkContactUsLinkComponent
LC-MS nitrogen generators
LC-MS nitrogen generators
Supplier:   PerkinElmer
LC-MS nitrogen generators
Supplier:  PerkinElmer
Certificates
LC-MS nitrogen generators
2 Options Available Below
Product Details & Documents
Nitrogen generators are designed to supply single or multiple LC-MS instruments with dry nitrogen at purities of 98 – 99,5%. The generator can also be used for solvent evaporation as well as supplying dry nitrogen to analytical instruments.
Product Family Options
Nitroflow lab
Product Information
  • Type
    Flow rate
    W×D×H
    Availability
    Price
  • Low Flow NitroFlow Lab, with air compressor
    10 - 30 l/min
    900×310×700 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 43 250,00 €
    43 250,00 €
    1 * 1 ST
       43 250,00 €
Generator without air compressor
Product Information
  • Type
    Flow rate
    W×D×H
    Availability
    Price
  • N2-35 Mid Flow, without air compressor
    20 l/min at 100 psi; 25 l/min at 125 psi; 30 l/min at 100 psi; 37 l/min at 125 psi; 46 l/min at 100 psi; 57 l/min at 125 psi
    450×410×1300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 42 210,00 €
    42 210,00 €
    1 * 1 ST
       42 210,00 €