Orders LinkContactUsLinkComponent
Safety spectacles, Adaptec™
Safety spectacles, Adaptec™
Supplier:   HONEYWELL SAFETY
Safety spectacles, Adaptec™
Supplier:  HONEYWELL SAFETY
Certificates
Safety spectacles, Adaptec™
Safety spectacles, Adaptec™
3 Options Available Below
Product Details & Documents
Stylish single lens safety glasses with vented, cushioned frame and PC lens.
  • Three position nose bridge is simple to adjust and keeps eyes centred in the lens
  • Self-adjusting nose pads
  • Adjustable lens angle reduces fogging; each temple hinge ratchets to seven positions
  • Temple microadjustment allows workers to adjust temple tension without removing eyewear
  • 4A+ coating on both sides of the lens ensures excellent anti-fog, anti-static and scratch resistant properties
Suitable for jobs where there are risks associated with abrasion, fogging, airborne particulates or impact.
Certifications: EN 166 FT
Product Family Options
Product Information
  • Type
    Colour
    Lenses
    Availability
    Price
  • Anti-scratch, anti-fog, anti-static (narrow version)
    Black/red
    Clear HS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 21,80 €
    21,80 €
    1 * 1 ST
       21,80 €
  • Anti-scratch, anti-fog, anti-static (regular version)
    Black/red
    Clear HS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 20,50 €
    20,50 €
    1 * 1 ST
       20,50 €
  • Anti-scratch, anti-fog, anti-static (wide version)
    Black/red
    Clear HS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 21,80 €
    21,80 €
    1 * 1 ST
       21,80 €