Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

pH buffers, DuraCal

pH buffers, DuraCal
Supplier:  HAMILTON BONADUZ
Certificates
pH buffers, DuraCal
pH buffers, DuraCal

18 Options Available Below

Product Details & Documents

For full information concerning health and safety data please contact Avantor or Hamilton directly.

  • Label includes expiration date, batch number and actual value
  • Traceable to NIST and PTB, certificates available to download
  • Stable solutions
  • Certified pH value from a DAkkS laboratory accredited for pH measurement
Product Family Options

Product Information

  • pH value
    pH accuracy
    pH buffer type
    Colour
    Pack type
    Pk Info
    Pk
    Availability
    Price
  • 2,00
    ±0,02
    Colourless
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 33,80 €
    33,80 €
    1 * 500 mL
       33,80 €

  • 4,01
    ±0,02
    Pink
    Plastic bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 24,50 €
    24,50 €
    1 * 250 mL
       24,50 €

  • 4,01
    ±0,02
    Pink
    Plastic bottle
    3×500 ml
    3
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 82,50 €
    82,50 €
    1 * 3 ST
       82,50 €

  • 4,01
    ±0,02
    Pink
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 165,00 €
    165,00 €
    1 * 5 L
       165,00 €

  • 4,01
    ±0,02
    Pink
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 282,00 €
    282,00 €
    1 * 10 L
       282,00 €

  • 5,00
    ±0,02
    Colourless
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 33,80 €
    33,80 €
    1 * 500 mL
       33,80 €

  • 7,00
    ±0,02
    Green
    Plastic bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 24,50 €
    24,50 €
    1 * 250 mL
       24,50 €

  • 7,00
    ±0,02
    Green
    Plastic bottle
    3×500 ml
    3
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 82,50 €
    82,50 €
    1 * 3 ST
       82,50 €

  • 7,00
    ±0,02
    Green
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 165,00 €
    165,00 €
    1 * 5 L
       165,00 €

  • 7,00
    ±0,02
    Green
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 295,00 €
    295,00 €
    1 * 10 L
       295,00 €

  • 8,00
    ±0,02
    Colourless
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 33,80 €
    33,80 €
    1 * 500 mL
       33,80 €

  • 9,21
    ±0,02
    Blue
    Plastic bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 24,50 €
    24,50 €
    1 * 250 mL
       24,50 €

  • 9,21
    ±0,02
    Blue
    Plastic bottle
    3×500 ml
    3
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 82,50 €
    82,50 €
    1 * 3 ST
       82,50 €

  • 10,01
    ±0,02
    Carbonate
    Yellow
    Plastic bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 24,50 €
    24,50 €
    1 * 250 mL
       24,50 €

  • 10,01
    ±0,02
    Carbonate
    Yellow
    Plastic bottle
    500 ml
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 31,00 €
    31,00 €
    1 * 500 mL
       31,00 €

  • 10,01
    ±0,02
    Carbonate
    Blue
    Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 282,00 €
    282,00 €
    1 * 10 L
       282,00 €

  • 11,00
    ±0,05
    Colourless
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 39,20 €
    39,20 €
    1 * 500 mL
       39,20 €

  • 12,00
    ±0,05
    Colourless
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 39,20 €
    39,20 €
    1 * 500 mL
       39,20 €