Orders LinkContactUsLinkComponent
Rectangular cover glasses
Rectangular cover glasses
Rectangular cover glasses
Certificates
Rectangular cover glasses
Rectangular cover glasses
11 Options Available Below
Product Details & Documents
Cover glass D 263® M made of borosilicate glass.
  • Free from colour distortion and highly transparent
  • Excellent quality, low levels of inclusions, striae, bubbles and streaks
  • High spectral transmission, excellent flatness, optimum resistance to chemicals
Certifications: ISO 8255
Product Family Options
Product Information
  • L×W
    Thickness
    Availability
    Price
  • 18×24 mm
    0,13 - 0,16 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 47,00 €
    47,00 €
    1 * 1.000 ST
       47,00 €
  • 22×32 mm
    0,13 - 0,16 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 68,60 €
    68,60 €
    1 * 1.000 ST
       68,60 €
  • 22×32 mm
    0,16 - 0,19 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST
  • 24×30 mm
    0,13 - 0,16 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 76,40 €
    76,40 €
    1 * 1.000 ST
       76,40 €
  • 24×32 mm
    0,13 - 0,16 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 64,10 €
    64,10 €
    1 * 1.000 ST
       64,10 €
  • 24×40 mm
    0,13 - 0,16 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 82,20 €
    82,20 €
    1 * 1.000 ST
       82,20 €
  • 24×50 mm
    0,13 - 0,16 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 79,10 €
    79,10 €
    1 * 1.000 ST
       79,10 €
  • 24×55 mm
    0,13 - 0,16 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 9,50 €
    9,50 €
    1 * 100 ST
       9,50 €
  • 24×60 mm
    0,13 - 0,16 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 90,50 €
    90,50 €
    1 * 1.000 ST
       90,50 €
  • 33×50 mm
    0,13 - 0,16 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 689,00 €
    689,00 €
    1 * 100 ST
       689,00 €
  • 45×60 mm
    0,13 - 0,16 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 307,00 €
    307,00 €
    1 * 1.000 ST
       307,00 €
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.