Diaphragm valves, VM series
Supplier: VACUUBRAND
Certificates
About this item
VM series valves are hand operated diaphragm valves. They are used as isolation valves, air-admittance valves or manual-control valves.
- Materials with high chemical resistance
- High conductance, low leakage rate
- Flow control possible
- Gas-tight against atmosphere in both directions
Frequently Bought Together