Orders LinkContactUsLinkComponent
Sample cups for auto analyser
Sample cups for auto analyser
Sample cups for auto analyser
Cups made of PS for auto analyser.
  • Good chemical resistance
  • Good temperature resistance
Product Family Options
  • Description
    Capacity
  • For Centrifichem autoanalysers
    0,25 ml
    1 * 1.000 ST
    1 * 1.000 ST
  • For Genseac autoanalysers
    0,5 ml
    1 * 1.000 ST
    1 * 1.000 ST
  • For Hitachi autoanalysers
    3,0 ml
    1 * 1.000 ST
    1 * 1.000 ST