Cylinders, without lid
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
About this item
Jars, made of glass or stainless steel, with base, without lids.
Suitable for storing dressing material, cotton, clinical thermometers, forceps and other instruments.
Ordering information: Lids with knob available must be ordered separately.
14 Options Available Below
Product Details & Documents
Jars, made of glass or stainless steel, with base, without lids.
Suitable for storing dressing material, cotton, clinical thermometers, forceps and other instruments.
Ordering information: Lids with knob available must be ordered separately.
Product Family Options
Glass
Product Information
- HeightExt.ØAvailabilityPrice
Stainless steel
Product Information
- HeightExt.ØAvailabilityPrice
Lids