Orders LinkContactUsLinkComponent
Ground joint reducing set, BOLA
Ground joint reducing set, BOLA
Reducing set, NS 14, NS 19, NS 24, NS 29, NS 34, NS 45 and NS 60
Catalog # BOHLH981-14
Supplier:   Bohlender
Ground joint reducing set, BOLA
Catalog # BOHLH981-14
Supplier:  Bohlender
Ground joint reducing set, BOLA
Ground joint reducing set, BOLA
Reducing set, NS 14, NS 19, NS 24, NS 29, NS 34, NS 45 and NS 60
Price
138,00 €
Log in for availability
1 * 1 SET
Product Details & Documents
Ground joint reducing set, PTFE, consisting of reducing rings graduated for bridging different cone and socket sizes.
  • Universal chemical resistance
  • Temperature resistance –200 to +250 °C
  • Suitable for vacuum applications
Specifications
  • For:
    NS 14, NS 19, NS 24, NS 29, NS 34, NS 45 and NS 60
  • Description:
    Reducing set