O-rings for laboratory flat flanges, BOLA
FEP. Sealing for flat flange with groove.
- Temperature resistance: from –60 to +205 °C
- Excellent chemical resistance
Silicone core with seamless FEP coating; manufactured according to DIN 12214;1996-12; flexible, almost universal chemical resistance.
Frequently Bought Together