Orders LinkContactUsLinkComponent
PIG® small mobile container kit
Supplier:   NEW PIG
PIG® small mobile container kit
Supplier:  NEW PIG
Certificates
PIG® small mobile container kit
PIG® small mobile container kit
6 Options Available Below
Product Details & Documents
Fast, effective response to indoor and outdoor spills.
  • Durable, wheeled 120 L container is packed with high-quality PIG® absorbents
  • Easy to transport supplies directly to the spill site
  • Hinged lid makes it easy to get the absorbents you need — just flip it open
  • Container with bold Spill Kit lettering for quick identification
  • Absorbents are organised for quick, easy response
  • Heavy-duty rubber wheels roll easily over nearly any surface
The weather-resistant polyethylene container protects the contents from moisture, dirt and damage (containers are best kept indoors for long-term storage).
 : To ensure effectiveness and your safety, we recommend that you conduct compatibility and absorption testing of your chemicals with PIG® HAZ-MAT products prior to purchase.
Product Family Options
Universal
Product Information
  • Absorbency
    W×D×H
    Availability
    Price
  • 121-0061
    Spill kit, includes 100 mat pads, 10 socks, 12 pillows and 5 disposal bags
    Spill kit, includes 100 mat pads, 10 socks, 12 pillows and 5 disposal bags
    87,9 L
    550×480×970 mm
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 228,00 €
    228,00 €
    1 * 1 KIT
       228,00 €
  • 121-0117
    Refill only
    Refill only
    87,9 L
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 125,00 €
    125,00 €
    1 * 1 ST
       125,00 €
HAZ-MAT
Product Information
  • Absorbency
    W×D×H
    Availability
    Price
  • 121-0067
    Spill kit, includes 100 mat pads, 8 socks, 12 pillows and 5 disposal bags
    Spill kit, includes 100 mat pads, 8 socks, 12 pillows and 5 disposal bags
    80,7 L
    550×480×970 mm
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 325,00 €
    325,00 €
    1 * 1 KIT
       325,00 €
  • 121-0122
    Refill only
    Refill only
    80,7 L
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 221,00 €
    221,00 €
    1 * 1 ST
       221,00 €
Oil-Only
Product Information
  • Absorbency
    W×D×H
    Availability
    Price
  • 121-0071
    Spill kit, includes 50 mat pads, 4 socks,2 booms, 4 pillows and 5 disposal bags
    Spill kit, includes 50 mat pads, 4 socks,2 booms, 4 pillows and 5 disposal bags
    79,6 L
    550×480×970 mm
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 249,00 €
    249,00 €
    1 * 1 KIT
       249,00 €
  • 121-0125
    Refill only
    Refill only
    79,6 L
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 144,00 €
    144,00 €
    1 * 1 ST
       144,00 €