3-Hydroxypyridine N-oxide
Catalog # MOLEM44132528
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:111.0987 g/mol
EINECS:229-545-4
CAS Number:6602-28-4
Formula:C₅H₅NO₂
MDL Number:MFCD00006200