Order Entry
Belgium
ContactUsLinkComponent
 
L(-)-Malic acid monosodium salt
Catalog # SERA11064.02
Supplier:  Serva
CAS Number:  
L(-)-Malic acid monosodium salt
Catalog # SERA11064.02
Supplier:  Serva
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
CAS Number:68303-40-2
Formula:C₄H₅NaO₅