Order Entry
Belgium
ContactUsLinkComponent
Ammonium nitrate 2%
Ammonium nitrate 2%
Catalog # CMPL909D.20K.W.L1
CAS Number:  
Ammonium nitrate 2%
Catalog # CMPL909D.20K.W.L1
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :80.04 g/mol
Storage Temperature:Ambient
EINECS:229-347-8
CAS Number:6484-52-2
Formula:NH₄NO₃
MDL Number:MFCD00011425