Order Entry
Belgium
ContactUsLinkComponent
 
Phenanthrene-[D10] 200 µg/ml in isooctane (98% D), CIL
  PROOCILDLM-371-1.2
 :  
Phenanthrene-[D10] 200 µg/ml in isooctane (98% D), CIL
  PROOCILDLM-371-1.2
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    1517-22-2
  • Description:
    Phenanthrene-[D10] 200 µg/ml in isooctane (98% D)
  • Formula:
    C₁₄D₁₀
  • Description:
    Phenanthrene-[D10]