

Acetonitrile ≥99.8%, SupraSolv® for GC-MS
: Merck
:75-05-8
:
:H₃CCN
:MFCD00001878
:41.05 g/mol
:13,00071
:81.6 °C (1013 hPa)
:–45.7 °C
:0.782 g/cm³ (20 °C)
:200-835-2
:1648
:3,II
:M-Clarity™ quality level = MQ100
:2 °C (closed cup)
:01-2119471307-38
:GHS02
:GHS07
:Ambient
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
- Purity (GC):=99.8%
- Identity(IR):Conforms
- Evaporation Residue:= 3.0 mg/l
- Water:= 0.05%
- Color:=10 Hazen
- GC/MSD (retention range n-undecane to n-tetracontane, scanning area 30-600 amu, individual signals (n-tetradecane standard)):= 3ng/ml
- Suitability:Residue Analysis
Frequently Bought Together