Order Entry
Belgium
Orders LinkContactUsLinkComponent
Drum dolly, adjustable

Drum dolly, adjustable

Adjustable drum dolly, for 20 l drums and containers Ø 57,5 - 67,5 mm, Black, 14,05 kg, 670×130 mm
Catalog # NEWPDLY301-01
Supplier:   NEW PIG
Drum dolly, adjustable
Catalog # NEWPDLY301-01
Supplier:  NEW PIG
Drum dolly, adjustable
Drum dolly, adjustable

Adjustable drum dolly, for 20 l drums and containers Ø 57,5 - 67,5 mm, Black, 14,05 kg, 670×130 mm

Price
237,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

This drum dolly with adjustable diameter quickly expands for a precise fit on drums or overpacks up to Ø 677 mm and 363 kg.

  • Heavy-duty 5 mm steel frame adjusts in minutes with bolts and lock nuts
  • Curved ends hold drum tight
  • Wide, stable wheelbase helps prevent tipovers
  • Hard rubber casters for smooth rolling
  • All four casters have brakes to offer secure positioning and keep dolly from moving

Wheel Ø 70 mm

Specifications
  • Weight:
    14,05 kg
  • Colour:
    Black
  • Description:
    Adjustable drum dolly, for 20 l drums and containers Ø 57,5 - 67,5 mm
  • Ø×H:
    670×130 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.