Orders LinkContactUsLinkComponent
Filtering flasks with glass hose connection
Filtering flasks with glass hose connection
Filtering flasks with glass hose connection
Certificates
Filtering flasks with glass hose connection
Filtering flasks with glass hose connection
4 Options Available Below
Product Details & Documents
Flasks for vacuum use, made of DURAN® borosilicate glass.
  • Erlenmeyer shape
  • Olive hose connection Ø 11 mm
  • Does not conform to the 'Equipment and product safety regulations'
ISO 6556
Product Family Options
Product Information
  • Capacity
    Availability
    Price
  • 250 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 66,30 €
    66,30 €
    1 * 1 ST
       66,30 €
  • 500 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 72,10 €
    72,10 €
    1 * 1 ST
       72,10 €
  • 1000 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 99,60 €
    99,60 €
    1 * 1 ST
       99,60 €
  • 2000 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 120,00 €
    120,00 €
    1 * 1 ST
       120,00 €