Orders LinkContactUsLinkComponent
Automatic burettes, Schilling pattern, PMMA, class B
Automatic burettes, Schilling pattern, PMMA, class B
Supplier:   witeg Labortechnik
Automatic burettes, Schilling pattern, PMMA, class B
Supplier:  witeg Labortechnik
Certificates
Automatic burettes, Schilling pattern, PMMA, class B
Automatic burettes, Schilling pattern, PMMA, class B
2 Options Available Below
Product Details & Documents
Automatic burettes, class DIN-B, short line graduation, clear plastic (PMMA).
  • Push button enabling a precise titration
  • Complete with polyethylene reservoir
  • Included with a stable base
  • Automatic zero point
Liquid delivery time 30 to 60 seconds, without waiting time.
Product Family Options
Product Information
  • Capacity
    Division
    Tolerance
    Availability
    Price
  • 25 ml
    0,1 ml
    0,04 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 113,00 €
    113,00 €
    1 * 1 ST
       113,00 €
  • 50 ml
    0,1 ml
    0,08 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 118,00 €
    118,00 €
    1 * 1 ST
       118,00 €
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.